ContactUsLinkComponent
qPCR detection of Zaire ebola virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome virus es only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-EBOV-
Supplier:  PRIMERDESIGN
qPCR detection of Zaire ebola virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome virus es only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-EBOV-
Supplier:  PRIMERDESIGN
Frequently Bought Together