ContactUsLinkComponent
qPCR detection of Epstein Barr Virus (HHV4). Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA geno me viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-EBV-S
Supplier:  PRIMERDESIGN
qPCR detection of Epstein Barr Virus (HHV4). Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA geno me viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-EBV-S
Supplier:  PRIMERDESIGN
Frequently Bought Together