ContactUsLinkComponent
qPCR detection of Canine Distemper Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-CDV-S
Supplier:  PRIMERDESIGN
qPCR detection of Canine Distemper Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-CDV-S
Supplier:  PRIMERDESIGN
Frequently Bought Together