ContactUsLinkComponent
qPCR detection of all Clostridium perfringens species. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-C.PER
Supplier:  PRIMERDESIGN
qPCR detection of all Clostridium perfringens species. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-C.PER
Supplier:  PRIMERDESIGN
Frequently Bought Together