qPCR detection of Capripoxvirus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses o nly), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-CAPV-
Supplier: PRIMERDESIGN
Frequently Bought Together