ContactUsLinkComponent
qPCR detection of Human Parainfluenza virus type 4B. Contains: Primer and probe mix, Reverse Transcription target specific primers ( RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HPI4B
Supplier:  PRIMERDESIGN
qPCR detection of Human Parainfluenza virus type 4B. Contains: Primer and probe mix, Reverse Transcription target specific primers ( RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HPI4B
Supplier:  PRIMERDESIGN
Frequently Bought Together