ContactUsLinkComponent
qPCR detection of Sheep. Contains: Primer and probe mix, Positive control template, Internal extraction control 1 * 100 Tests
Catalog # PATH-SS-O.ARIE
Supplier:  PRIMERDESIGN
qPCR detection of Sheep. Contains: Primer and probe mix, Positive control template, Internal extraction control 1 * 100 Tests
Catalog # PATH-SS-O.ARIE
Supplier:  PRIMERDESIGN
Frequently Bought Together