ContactUsLinkComponent
qPCR detection of Chicken. Contains: Primer and probe mix, Positive control template, Internal extraction control 1 * 100 Tests
Catalog # PATH-SS-G.GALL
Supplier:  PRIMERDESIGN
qPCR detection of Chicken. Contains: Primer and probe mix, Positive control template, Internal extraction control 1 * 100 Tests
Catalog # PATH-SS-G.GALL
Supplier:  PRIMERDESIGN
Frequently Bought Together