ContactUsLinkComponent
qPCR detection of Human Rhinovirus Subtype 9. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard cu rve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HRV9
Supplier:  PRIMERDESIGN
qPCR detection of Human Rhinovirus Subtype 9. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard cu rve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HRV9
Supplier:  PRIMERDESIGN
Frequently Bought Together