ContactUsLinkComponent
qPCR detection of Methicillin-resistant Staphylococcus aureus with Staphylococcal cassette chromosome mec(SCC mec). Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-MRSA
Supplier:  PRIMERDESIGN
qPCR detection of Methicillin-resistant Staphylococcus aureus with Staphylococcal cassette chromosome mec(SCC mec). Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-MRSA
Supplier:  PRIMERDESIGN
Frequently Bought Together