ContactUsLinkComponent
qPCR detection of Methicillin-resistant Staphylococcus aureus with Staphylococcal cassette chromosome mec(SCC mec) type IVa. Contain s: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standar 1 * 150 Tests
Catalog # PATH-STD-MRSA4
Supplier:  PRIMERDESIGN
qPCR detection of Methicillin-resistant Staphylococcus aureus with Staphylococcal cassette chromosome mec(SCC mec) type IVa. Contain s: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standar 1 * 150 Tests
Catalog # PATH-STD-MRSA4
Supplier:  PRIMERDESIGN
Frequently Bought Together