ContactUsLinkComponent
qPCR detection of Human Rhinovirus Subtype 1B. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA ge nome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HRV1B
Supplier:  PRIMERDESIGN
qPCR detection of Human Rhinovirus Subtype 1B. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA ge nome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HRV1B
Supplier:  PRIMERDESIGN
Frequently Bought Together