ContactUsLinkComponent
qPCR detection of Human T-lymphotropic virus Type I. Contains: Primer and probe mix, Reverse Transcription target specific primers ( RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HTLV1
Supplier:  PRIMERDESIGN
qPCR detection of Human T-lymphotropic virus Type I. Contains: Primer and probe mix, Reverse Transcription target specific primers ( RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HTLV1
Supplier:  PRIMERDESIGN
Frequently Bought Together