ContactUsLinkComponent
qPCR detection of Mycobacterium avium. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome vir uses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-M.AVI
Supplier:  PRIMERDESIGN
qPCR detection of Mycobacterium avium. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome vir uses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-M.AVI
Supplier:  PRIMERDESIGN
Frequently Bought Together