qPCR detection of Israeli Acute Paralysis Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-IAPV
Supplier: PRIMERDESIGN
Frequently Bought Together