ContactUsLinkComponent
qPCR detection of Streptococcus sanguinis. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
  PATH-STD-S.SAN
qPCR detection of Streptococcus sanguinis. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
  PATH-STD-S.SAN
Frequently Bought Together