qPCR detection of Spring Viremia of Carp Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA g enome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-SVCV-
Supplier: PRIMERDESIGN
Frequently Bought Together