ContactUsLinkComponent
qPCR detection of Avian Influenza A Virus Subtype H9. Contains: Primer and probe, RT target specific primers (RNA viruses only), Sta ndard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-H9
Dodavatel:  PRIMERDESIGN
qPCR detection of Avian Influenza A Virus Subtype H9. Contains: Primer and probe, RT target specific primers (RNA viruses only), Sta ndard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-H9
Dodavatel:  PRIMERDESIGN
Frequently Bought Together