ContactUsLinkComponent
qPCR detection of Peste-des-petits-ruminants Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Stand ard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-PPRV
Supplier:  PRIMERDESIGN
qPCR detection of Peste-des-petits-ruminants Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Stand ard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-PPRV
Supplier:  PRIMERDESIGN
Frequently Bought Together