ContactUsLinkComponent
qPCR detection of Mycoplasma pneumoniae. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-M.PNEUMON
Supplier:  PRIMERDESIGN
qPCR detection of Mycoplasma pneumoniae. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-M.PNEUMON
Supplier:  PRIMERDESIGN
Frequently Bought Together