ContactUsLinkComponent
qPCR detection of Plasmodium vivax. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruse s only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-P.VIV
Supplier:  PRIMERDESIGN
qPCR detection of Plasmodium vivax. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruse s only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-P.VIV
Supplier:  PRIMERDESIGN
Frequently Bought Together