ContactUsLinkComponent
qPCR detection of Peste-des-petits-ruminants Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (R NA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-PPRV-
Supplier:  PRIMERDESIGN
qPCR detection of Peste-des-petits-ruminants Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (R NA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-PPRV-
Supplier:  PRIMERDESIGN
Frequently Bought Together