ContactUsLinkComponent
qPCR detection of Mycoplasma hyopneumoniae. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curv e, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-M.HYOPNEU
Supplier:  PRIMERDESIGN
qPCR detection of Mycoplasma hyopneumoniae. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curv e, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-M.HYOPNEU
Supplier:  PRIMERDESIGN
Frequently Bought Together