ContactUsLinkComponent
qPCR detection of Avian adenovirus EDS76 Egg Drop Syndrome. Contains: Primer and probe, RT target specific primers (RNA viruses only ), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-EDS
Supplier:  PRIMERDESIGN
qPCR detection of Avian adenovirus EDS76 Egg Drop Syndrome. Contains: Primer and probe, RT target specific primers (RNA viruses only ), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-EDS
Supplier:  PRIMERDESIGN
Frequently Bought Together