qPCR detection of Haemophilus ducreyi. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, En dogenous control, extraction control 1 * 150 Tests
Catalog # PATH-H.DUCREYI
Supplier: PRIMERDESIGN
Frequently Bought Together