ContactUsLinkComponent
qPCR detection of Dengue virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenou s control, extraction control 1 * 150 Tests
Catalog # Z-PATH-DENG
Supplier:  PRIMERDESIGN
qPCR detection of Dengue virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenou s control, extraction control 1 * 150 Tests
Catalog # Z-PATH-DENG
Supplier:  PRIMERDESIGN
Frequently Bought Together