ContactUsLinkComponent
qPCR detection of Campylobacter Coli. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, End ogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-C.COLI
Supplier:  PRIMERDESIGN
qPCR detection of Campylobacter Coli. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, End ogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-C.COLI
Supplier:  PRIMERDESIGN
Frequently Bought Together