ContactUsLinkComponent
qPCR detection of Bluetongue Virus 1. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, End ogenous control, extraction control 1 * 150 Tests
Catalog # PATH-BTV-1
Supplier:  PRIMERDESIGN
qPCR detection of Bluetongue Virus 1. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, End ogenous control, extraction control 1 * 150 Tests
Catalog # PATH-BTV-1
Supplier:  PRIMERDESIGN
Frequently Bought Together