ContactUsLinkComponent
Real-time PCR Genotyping kit to quantify BRAF (V600E). Contains: Primer and multiplex probe mix - wildtype/variants, Positive contro ls - wildtype/variants, quasa mastermix 1 * 50 Tests
Catalog # PATH-GT-QU-BRA
Supplier:  PRIMERDESIGN
Real-time PCR Genotyping kit to quantify BRAF (V600E). Contains: Primer and multiplex probe mix - wildtype/variants, Positive contro ls - wildtype/variants, quasa mastermix 1 * 50 Tests
Catalog # PATH-GT-QU-BRA
Supplier:  PRIMERDESIGN
Frequently Bought Together