ContactUsLinkComponent
qPCR detection of Rift Valley Fever Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve , Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-RVFV
Supplier:  PRIMERDESIGN
qPCR detection of Rift Valley Fever Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve , Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-RVFV
Supplier:  PRIMERDESIGN
Frequently Bought Together