ContactUsLinkComponent
qPCR detection of Borrelia burgdorferi. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, E ndogenous control, extraction control 1 * 150 Tests
Catalog # PATH-B.BURGDOR
Supplier:  PRIMERDESIGN
qPCR detection of Borrelia burgdorferi. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, E ndogenous control, extraction control 1 * 150 Tests
Catalog # PATH-B.BURGDOR
Supplier:  PRIMERDESIGN
Frequently Bought Together