ContactUsLinkComponent
Real-time PCR Genotyping kit to detect C2 codon 318. Contains: Primer and multiplex probe mix - wildtype/variants, Positive controls - wildtype/variants 1 * 50 Tests
Catalog # PATH-GT-C2-318
Supplier:  PRIMERDESIGN
Real-time PCR Genotyping kit to detect C2 codon 318. Contains: Primer and multiplex probe mix - wildtype/variants, Positive controls - wildtype/variants 1 * 50 Tests
Catalog # PATH-GT-C2-318
Supplier:  PRIMERDESIGN
Frequently Bought Together