ContactUsLinkComponent
GMO Figwort Mosaic Virus screening kit. Contains: Target specific primer/probe mix, Wild-type virus primer/probe mix, Internal extra ction control, Positive control 1 * 100 Tests
Catalog # PATH-GMO-FMV
Supplier:  PRIMERDESIGN
GMO Figwort Mosaic Virus screening kit. Contains: Target specific primer/probe mix, Wild-type virus primer/probe mix, Internal extra ction control, Positive control 1 * 100 Tests
Catalog # PATH-GMO-FMV
Supplier:  PRIMERDESIGN
Frequently Bought Together