ContactUsLinkComponent
General GMO screening kit for 35S promoter in GM crops. Contains: Target specific primer/probe mix, Wild-type virus primer/probe mix , Internal extraction control, Positive control 1 * 100 Tests
Catalog # PATH-GMO-P35S
Supplier:  PRIMERDESIGN
General GMO screening kit for 35S promoter in GM crops. Contains: Target specific primer/probe mix, Wild-type virus primer/probe mix , Internal extraction control, Positive control 1 * 100 Tests
Catalog # PATH-GMO-P35S
Supplier:  PRIMERDESIGN
Frequently Bought Together