ContactUsLinkComponent
qPCR detection of Bovine herpesvirus 1. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, E ndogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-BHV-1
Supplier:  PRIMERDESIGN
qPCR detection of Bovine herpesvirus 1. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, E ndogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-BHV-1
Supplier:  PRIMERDESIGN
Frequently Bought Together