ContactUsLinkComponent
qPCR detection of all species within the Burkholderia cepacia complex. Contains: Primer and probe, RT target specific primers (RNA v iruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-B.CEPACIA
Supplier:  PRIMERDESIGN
qPCR detection of all species within the Burkholderia cepacia complex. Contains: Primer and probe, RT target specific primers (RNA v iruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-B.CEPACIA
Supplier:  PRIMERDESIGN
Frequently Bought Together