ContactUsLinkComponent
IQ/OQ document with works test data for chamber, incl. Temperature distribution survey for one free-selectable temperature value. 9 measuring points (size30), 27 measuring points (size 55 - 1060) to DIN 12 880: 2007-05 (further temperature values on 1 * 1 items
Catalog # MEMMD00127UF1060
Supplier:  MEMMERT
IQ/OQ document with works test data for chamber, incl. Temperature distribution survey for one free-selectable temperature value. 9 measuring points (size30), 27 measuring points (size 55 - 1060) to DIN 12 880: 2007-05 (further temperature values on 1 * 1 items
Catalog # MEMMD00127UF1060
Supplier:  MEMMERT
Frequently Bought Together