ContactUsLinkComponent
Potential-free contact (24 V/2 A) with socket to NAMUR NE 28, for signal generation, controlled by programme segment, for feely sele ctable functions to be activated (e.g. activation of audible an visual signals, exhaust motors, fans, stirrers, etc.) 1 * 1 items
Catalog # MEMMUF1060PLUSH72
Supplier:  MEMMERT
Potential-free contact (24 V/2 A) with socket to NAMUR NE 28, for signal generation, controlled by programme segment, for feely sele ctable functions to be activated (e.g. activation of audible an visual signals, exhaust motors, fans, stirrers, etc.) 1 * 1 items
Catalog # MEMMUF1060PLUSH72
Supplier:  MEMMERT
Frequently Bought Together