ContactUsLinkComponent
Fan speed monitoring with switching off the heating and with alarm in case of failure - optional for Ufplus only 1 * 1 items
Catalog # MEMMUF1060PLUSV4
Supplier:  MEMMERT
Fan speed monitoring with switching off the heating and with alarm in case of failure - optional for Ufplus only 1 * 1 items
Catalog # MEMMUF1060PLUSV4
Supplier:  MEMMERT
Frequently Bought Together