ContactUsLinkComponent
IQ/OQ document with works test data for chamber, incl. Temperature distribution survey for one free-selectable temperature, humidity and light value. 27 measuring points to DIN 12 880: 2007-05, PQ check list as support for validation by customer (mo 1 * 1 items
Catalog # MEMMD00137ICH110L
Supplier:  MEMMERT
IQ/OQ document with works test data for chamber, incl. Temperature distribution survey for one free-selectable temperature, humidity and light value. 27 measuring points to DIN 12 880: 2007-05, PQ check list as support for validation by customer (mo 1 * 1 items
Catalog # MEMMD00137ICH110L
Supplier:  MEMMERT
Frequently Bought Together