ContactUsLinkComponent
Extended overtemperature protection by additionally integrated Pt100 sensor for independent temperature monitoring for models UN/UF 1 * 1 items
Catalog # MEMMIF110A6
Supplier:  MEMMERT
Extended overtemperature protection by additionally integrated Pt100 sensor for independent temperature monitoring for models UN/UF 1 * 1 items
Catalog # MEMMIF110A6
Supplier:  MEMMERT
Frequently Bought Together