ContactUsLinkComponent
PRODUCT FOR DNA RELATE APPLICATION, 100 BASE-PAIR DNA LADDER, EXPRESS, CONSISTS OF MULTIPLE REPEATS OF 100-BP FRAGMENTS FROM 100 BP UP TO 1000 BP. SUPPLIED IN SUFFICIENT QUANTITY FOR 100 LOADINGS USING 4-5 UL PER LOADING. NOT DESIGNED FOR QUANTITATIN 1 * 500 µl
Catalog # ABNOD0028
Supplier:  Abnova
PRODUCT FOR DNA RELATE APPLICATION, 100 BASE-PAIR DNA LADDER, EXPRESS, CONSISTS OF MULTIPLE REPEATS OF 100-BP FRAGMENTS FROM 100 BP UP TO 1000 BP. SUPPLIED IN SUFFICIENT QUANTITY FOR 100 LOADINGS USING 4-5 UL PER LOADING. NOT DESIGNED FOR QUANTITATIN 1 * 500 µl
Catalog # ABNOD0028
Supplier:  Abnova
Frequently Bought Together