ContactUsLinkComponent
EC HPLC column EC 50/2 NUCLEOSIL 100-5 C18 AB Length: 50 mm, ID: 2 mm pack of 1 1 * 1 items
  MANA720957.20
EC HPLC column EC 50/2 NUCLEOSIL 100-5 C18 AB Length: 50 mm, ID: 2 mm pack of 1 1 * 1 items
  MANA720957.20
Frequently Bought Together