ContactUsLinkComponent
Standard rack for upright freezer for (hxd) 5x2 = 10 boxes 50 mm high, delivered including standard white cardboard boxes and 9x9 cell dividers, stainless steel, measures (hxdxw) 276 x 283 x 139mm 1 * 1 items
Catalog # TENATE23118B
Supplier:  TENAK
Standard rack for upright freezer for (hxd) 5x2 = 10 boxes 50 mm high, delivered including standard white cardboard boxes and 9x9 cell dividers, stainless steel, measures (hxdxw) 276 x 283 x 139mm 1 * 1 items
Catalog # TENATE23118B
Supplier:  TENAK
Frequently Bought Together