TECHNOVIT 9100 - COMPOUND 4: HARDENER 2 1 * 10 g
HKUL12225.00010
Frequently Bought Together
VWR®, Test strips, chloride, Dosatest
: VWR Chemicals
UltraClear® for histology, J.T.Baker®
: Avantor
NANOCOLOR® Photometric Tube Tests Kits for Ammonium
: MACHEREY-NAGEL
Formalin 10%, Q Path®, buffered
: VWR Chemicals
Ascorbic acid test strips, Dosatest
: VWR Chemicals
VWR®, OCT Embedding matrix for frozen sections
: VWR Chemicals
Test strips, peracetic acid, QUANTOFIX®
: MACHEREY-NAGEL
Ammonia solution ∼32%, GPR RECTAPUR®
: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Accessories for Technovit® 9100
: HERAEUS KULZER