Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Triamcinolone acetonide, 98% 1 * 250 mg
Catalog # J63548.MD
Supplier:
Thermo Fisher Scientific
SDS
Certificates
Expand image
Triamcinolone acetonide, 98% 1 * 250 mg
Catalog # J63548.MD
Supplier:
Thermo Fisher Scientific
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ63548
Application Notes
Organic Building Blocks and Intermediates
Documentation
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Formaldehyde ≥37% in aqueous solution stabilised, GR ACS, Reag. Ph. Eur. analytical reagent, Sigma-Aldrich®
Supplier: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Microscope Slides
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals