ContactUsLinkComponent
Blautouch - Interactive worktop 1800x750x900, Frame U , Tempered Glass 8mm (Black), Stainless Steel 304 + Windows +Scanner for bar code reading 2D 1 * 1 items
Catalog # LBRLBZ1ACAEABEA0BC
Supplier:  LABORIAL
Blautouch - Interactive worktop 1800x750x900, Frame U , Tempered Glass 8mm (Black), Stainless Steel 304 + Windows +Scanner for bar code reading 2D 1 * 1 items
Catalog # LBRLBZ1ACAEABEA0BC
Supplier:  LABORIAL
Frequently Bought Together