ContactUsLinkComponent
CXCL1 Rat Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 73 amino acids and having a mole cular mass of 7.8 kDa. The Rat CXCL1 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80130-5
Supplier:  Biorbyt
CXCL1 Rat Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 73 amino acids and having a mole cular mass of 7.8 kDa. The Rat CXCL1 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80130-5
Supplier:  Biorbyt