CXCL13 Human Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 87 amino acids and having a m olecular mass of 10.3 kDa. The BCA-1 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80118-5
Supplier: Biorbyt
Frequently Bought Together