ContactUsLinkComponent
TFF3 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 69 amino acids (22-80 a.a.) wh ich includes 10 amino acid His Tag fused at N-terminus and having total molecular mass of 7.82 kDa. TFF3 Hum 1 * 5 µG
Catalog # BIRBORB80106-5
Supplier:  Biorbyt
TFF3 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 69 amino acids (22-80 a.a.) wh ich includes 10 amino acid His Tag fused at N-terminus and having total molecular mass of 7.82 kDa. TFF3 Hum 1 * 5 µG
Catalog # BIRBORB80106-5
Supplier:  Biorbyt